Please login to your online account to display your discounted pricing

5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  TG00105DA

 View more versions of this product

Catalog No. TG00105DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 224.71
Quantity 1g
Chemical Name or Material 5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol
Assay Percent Range 97%
CAS 17452-31-2
Packaging Amber Glass Bottle
Molecular Formula C10H9ClN2S

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol
Assay Percent Range 97%
Molecular Formula C10H9ClN2S
Formula Weight 224.71