Please login to your online account to display your discounted pricing

5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-(4-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrN2O
Formula Weight 239.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC50910CB

 
fisher scientific company
CC50910CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC50910DE

 
fisher scientific company
CC50910DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.07
Quantity 250mg
Assay 97%
Chemical Name or Material 5-(4-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Assay Percent Range 97%
Molecular Formula C9H7BrN2O
CAS 71566-07-9
Packaging Amber Glass Bottle