Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Bromophenyl)-1H-tetrazole, 97%

$120.53 - $120.53

Chemical Identifiers

Chemical Name or Material 5-(4-Bromophenyl)-1H-tetrazole
CAS 50907-23-8
Molecular Formula C7H5BrN4
Formula Weight 225.05
MDL Number MFCD00991874
Beilstein 141235
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1518103

  SDS
alfa aesar™
L1518103
1g Each for $120.53
Description & Specifications

Specifications

Formula Weight 225.05
Chemical Name or Material 5-(4-Bromophenyl)-1H-tetrazole
Assay 97%
Molecular Formula C7H5BrN4
MDL Number MFCD00991874
CAS 50907-23-8
Melting Point ≈268°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 141235
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.