missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-[4′-(Bromomethyl)-1,1′-biphenyl-2-yl]-1-triphenylmethyl-1H-tetrazole, TCI America™

$143.61 - $429.31

Chemical Identifiers

CAS 124750-51-2
Molecular Formula C33H25BrN4
Molecular Weight (g/mol) 557.495
MDL Number MFCD00665221
InChI Key ZTFVTXDWDFIQEU-UHFFFAOYSA-N
Synonym 5-[4′C-(Bromomethyl)-1,1′C-biphenyl-2-yl]-1-trityl-1H-tetrazole
PubChem CID 10053461
IUPAC Name 5-[2-[4-(bromomethyl)phenyl]phenyl]-1-trityltetrazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C(=NN=N4)C5=CC=CC=C5C6=CC=C(C=C6)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44621G View Documents TCI America
B44621G
1g Each for $143.61
 
B44625G View Documents TCI America
B44625G
5g Each for $429.31
 
Specifications

Chemical Identifiers

124750-51-2
557.495
ZTFVTXDWDFIQEU-UHFFFAOYSA-N
10053461
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C(=NN=N4)C5=CC=CC=C5C6=CC=C(C=C6)CBr
C33H25BrN4
MFCD00665221
5-[4′C-(Bromomethyl)-1,1′C-biphenyl-2-yl]-1-trityl-1H-tetrazole
5-[2-[4-(bromomethyl)phenyl]phenyl]-1-trityltetrazole

Specifications

61°C
147°C
124750-51-2
MFCD00665221
5-[4′C-(Bromomethyl)-1,1′C-biphenyl-2-yl]-1-trityl-1H-tetrazole
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C(=NN=N4)C5=CC=CC=C5C6=CC=C(C=C6)CBr
557.495
557.50
≥98.0% (HPLC,T)
White
1g
C33H25BrN4
2811
ZTFVTXDWDFIQEU-UHFFFAOYSA-N
5-[2-[4-(bromomethyl)phenyl]phenyl]-1-trityltetrazole
10053461
Crystal-Powder at 20°C
5-[4′-(Bromomethyl)-1,1′-biphenyl-2-yl]-1-triphenylmethyl-1H-tetrazole
Safety and Handling

Safety and Handling

TSCA : No