missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Bromo-3-methylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5451903

 View more versions of this product

Catalog No. AAH5451903



Specifications

Specifications

5-(4-Bromo-3-methylphenylcarbamoyl)-3-fluorobenzeneboronic acid
97%
MFCD20040212
DIXOWPIMCKUEGL-UHFFFAOYSA-N
73995851
2096337-08-3
C14H12BBrFNO3
1g
351.97
351.97

Chemical Identifiers

2096337-08-3
351.97
DIXOWPIMCKUEGL-UHFFFAOYSA-N
C14H12BBrFNO3
MFCD20040212
73995851
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents