missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine, ≥97%, Maybridge

Chemical Identifiers

CAS 250726-93-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H17BO4S
Formula Weight 268.14
MDL Number MFCD08060541
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC47839DA SDS Fisher Scientific Company
CC47839DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC47839CB SDS Fisher Scientific Company
CC47839CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

250726-93-3
100.0
C12H17BO4S
MFCD08060541
95%
94.5
95%
268.14
5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine

Specifications

268.14
100.0
95%
94.5
5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine
1g
250726-93-3
C12H17BO4S
MFCD08060541
95%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning