missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, TCI America™

$187.92 - $583.63

Chemical Identifiers

CAS 269410-24-4
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.113
MDL Number MFCD03789263
InChI Key FATPQDPUKVVCLT-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, 1h-indole-5-boronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, indolyl-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester
PubChem CID 2734772
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05901G View Documents TCI America
I05901G
1g Each for $187.92
 
I05905G View Documents TCI America
I05905G
5g Each for $583.63
 
Specifications

Chemical Identifiers

269410-24-4
243.113
FATPQDPUKVVCLT-UHFFFAOYSA-N
2734772
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3
C14H18BNO2
MFCD03789263
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, 1h-indole-5-boronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, indolyl-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

Specifications

Undesignated
1g
C14H18BNO2
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, 1h-indole-5-boronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, indolyl-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3
243.113
243.11
≥98.0% (GC)
126°C
269410-24-4
MFCD03789263
FATPQDPUKVVCLT-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
2734772
Crystal-Powder at 20°C
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Safety and Handling

Safety and Handling

TSCA : No