missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 97+%, ACROS Organics™
Chemical Identifiers
CAS | 269410-24-4 |
---|---|
Molecular Formula | C14H18BNO2 |
Molecular Weight (g/mol) | 243.11 |
MDL Number | MFCD03789263 |
InChI Key | FATPQDPUKVVCLT-UHFFFAOYSA-N |
Synonym | 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1h-indole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester, indolyl-5-boronic acid pinacol ester |
PubChem CID | 2734772 |
IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC371782500
|
Thermo Fisher Scientific
AC371782500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC371780010
|
Thermo Fisher Scientific
AC371780010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
269410-24-4 | |
243.11 | |
FATPQDPUKVVCLT-UHFFFAOYSA-N | |
2734772 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3 |
C14H18BNO2 | |
MFCD03789263 | |
1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1h-indole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester, indolyl-5-boronic acid pinacol ester | |
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole |
Specifications
Authentic | |
Brown to Yellow | |
250mg | |
97.0 | |
97% min. (GC) | |
MFCD03789263 | |
Solubility in water: insoluble | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC=C3 | |
243.11 | |
243.11 | |
97+% |
Glass bottle | |
114°C to 116.5°C | |
269410-24-4 | |
100.0 | |
C14H18BNO2 | |
1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1h-indole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester, indolyl-5-boronic acid pinacol ester | |
FATPQDPUKVVCLT-UHFFFAOYSA-N | |
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole | |
2734772 | |
Crystalline Powder | |
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning