Please login to your online account to display your discounted pricing

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 97+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371780010

 View more versions of this product

Catalog No. AC371780010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Crystalline Powder
Formula Weight 243.11
Quantity 1g
Color Pale yellow to brown
Melting Point 124° to 128°C
CAS Min % 97.0
CAS Max % 100.0
MDL Number MFCD03789263
Synonym Indole-5-boronic acid pinacol ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Assay Percent Range 97+%
Molecular Formula C14H18BNO2
CAS 269410-24-4
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
CAS 269410-24-4
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C14H18BNO2
Formula Weight 243.11
MDL Number MFCD03789263
Synonym Indole-5-boronic acid pinacol ester
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents