missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde, TCI America™

Manufacturer:  TCI America C27865G

 View more versions of this product

Catalog No. C27865G



Specifications

Specifications

5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde
229°C
231278-84-5
MFCD10565688
C1=CC(=CC(=C1)F)COC2=C(C=C(C=C2)NC3=NC=NC4=C3C=C(C=C4)C5=CC=C(O5)C=O)Cl
473.888
473.89
≥97.0% (HPLC,T)
Yellow
5g
C26H17ClFN3O3
XQPZOUAAXRXPAM-UHFFFAOYSA-N
5-[4-[3-chloro-4-[(3-fluorophenyl)methoxy]anilino]quinazolin-6-yl]furan-2-carbaldehyde
11181296
Crystal-Powder at 20°C

Chemical Identifiers

231278-84-5
473.888
XQPZOUAAXRXPAM-UHFFFAOYSA-N
5-[4-[3-chloro-4-[(3-fluorophenyl)methoxy]anilino]quinazolin-6-yl]furan-2-carbaldehyde
C26H17ClFN3O3
MFCD10565688
11181296
C1=CC(=CC(=C1)F)COC2=C(C=C(C=C2)NC3=NC=NC4=C3C=C(C=C4)C5=CC=C(O5)C=O)Cl
Safety and Handling

Safety and Handling

TSCA : No

Documents