missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Trifluoromethoxyphenyl)-1H-tetrazole, 97%

Manufacturer:  Alfa Aesar™ L15268MC

 View more versions of this product

Catalog No. AAL15268MC



Specifications

Specifications

5-(3-Trifluoromethoxyphenyl)-1H-tetrazole
100mg
C8H5F3N4O
5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2
230.15
230.15
156°C to 159°C
216144-09-1
MFCD01075683
YKYWVXHVSVSQOX-UHFFFAOYSA-N
5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole
2777350
97%

Chemical Identifiers

216144-09-1
230.15
YKYWVXHVSVSQOX-UHFFFAOYSA-N
2777350
C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2
C8H5F3N4O
MFCD01075683
5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents