Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Pyridyl)-1H-tetrazole, 98%

$452.55 - $452.55

Chemical Identifiers

Chemical Name or Material 5-(3-Pyridyl)-1H-tetrazole
CAS 3250-74-6
Molecular Formula C6H5N5
Formula Weight 147.14
MDL Number MFCD00040281
Synonym 3-[1(2)H-Tetrazol-5-yl]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049206

 
alfa aesar™
B2049206
5g Each for $452.55
Description & Specifications

Specifications

Formula Weight 147.14
Chemical Name or Material 5-(3-Pyridyl)-1H-tetrazole
Assay 98%
Molecular Formula C6H5N5
MDL Number MFCD00040281
CAS 3250-74-6
Synonym 3-[1(2)H-Tetrazol-5-yl]pyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280d-P305+P351+P338
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Melting Point ≈240°C (dec.)
Quantity 5g

Applications
5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280d-P305+P351+P338
Wear protective clothing and eye protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H318
Causes serious eye damage.