missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Phenoxyphenyl)-1H-tetrazole, 98%

$53.97 - $151.76

Chemical Identifiers

CAS 374538-02-0
Molecular Formula C13H10N4O
Molecular Weight (g/mol) 238.25
MDL Number MFCD02093975
InChI Key TYNITQBHBADQAR-UHFFFAOYSA-N
Synonym 5-3-phenoxyphenyl-1h-tetrazole, 5-3-phenoxyphenyl-2h-tetrazole, 5-3-phenoxyphenyl-1h-1,2,3,4-tetrazole, acmc-1ctsy, 2h-tetrazole,5-3-phenoxyphenyl, 3-1h-1,2,3,4-tetraazol-5-yl-1-phenoxybenzene, 5-3-phenoxyphenyl-2h-1,2,3,4-tetrazole
PubChem CID 7010320
IUPAC Name 5-(3-phenoxyphenyl)-2H-tetrazole
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=NNN=N3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1795503 View Documents Alfa Aesar™
L1795503
1g Each for $151.76
 
AAL17955MD View Documents Alfa Aesar™
L17955MD
250mg Each for $53.97
 
Specifications

Chemical Identifiers

374538-02-0
238.25
TYNITQBHBADQAR-UHFFFAOYSA-N
7010320
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=NNN=N3
C13H10N4O
MFCD02093975
5-3-phenoxyphenyl-1h-tetrazole, 5-3-phenoxyphenyl-2h-tetrazole, 5-3-phenoxyphenyl-1h-1,2,3,4-tetrazole, acmc-1ctsy, 2h-tetrazole,5-3-phenoxyphenyl, 3-1h-1,2,3,4-tetraazol-5-yl-1-phenoxybenzene, 5-3-phenoxyphenyl-2h-1,2,3,4-tetrazole
5-(3-phenoxyphenyl)-2H-tetrazole

Specifications

152°C to 154°C
374538-02-0
MFCD02093975
TYNITQBHBADQAR-UHFFFAOYSA-N
5-(3-phenoxyphenyl)-2H-tetrazole
7010320
98%
1g
C13H10N4O
5-3-phenoxyphenyl-1h-tetrazole, 5-3-phenoxyphenyl-2h-tetrazole, 5-3-phenoxyphenyl-1h-1,2,3,4-tetrazole, acmc-1ctsy, 2h-tetrazole,5-3-phenoxyphenyl, 3-1h-1,2,3,4-tetraazol-5-yl-1-phenoxybenzene, 5-3-phenoxyphenyl-2h-1,2,3,4-tetrazole
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=NNN=N3
238.25
238.25
5-(3-Phenoxyphenyl)-1H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning