Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Methoxyphenyl)-1H-tetrazole, 98+%

$230.89 - $230.89

Chemical Identifiers

Chemical Name or Material 5-(3-Methoxyphenyl)-1H-tetrazole
CAS 73096-36-3
Molecular Formula C8H8N4O
Formula Weight 176.18
MDL Number MFCD03093094
Beilstein 1212228
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1926103

  SDS
alfa aesar™
L1926103
1g Each for $230.89
Description & Specifications

Specifications

Formula Weight 176.18
Assay ≥98%
Molecular Formula C8H8N4O
MDL Number MFCD03093094
CAS 73096-36-3
Beilstein 1212228
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-(3-Methoxyphenyl)-1H-tetrazole
Melting Point 156° to 158°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.