Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Fluorophenyl)-1H-tetrazole, 97%

$179.31 - $179.31

Chemical Identifiers

Chemical Name or Material 5-(3-Fluorophenyl)-1H-tetrazole
CAS 50907-20-5
Molecular Formula C7H5FN4
Formula Weight 164.14
MDL Number MFCD01075702
Beilstein 1104440
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1555803

  SDS
alfa aesar™
L1555803
1g Each for $179.31
Description & Specifications

Specifications

Formula Weight 164.14
MDL Number MFCD01075702
Chemical Name or Material 5-(3-Fluorophenyl)-1H-tetrazole
Health Hazard 1 Warning
CAS 50907-20-5
Melting Point 142° to 144°C
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H5FN4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Beilstein 1104440
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.