Learn More
5-(3-Chlorophenyl)-2-furoyl chloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H50404MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-(3-Chlorophenyl)-2-furoyl chloride | |
C11H6Cl2O2 | |
250 mg | |
5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl | |
ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1 | |
241.07 | |
241.08 |
92973-26-7 | |
MFCD02258034 | |
UN3261 | |
DBONYOXUBWINEQ-UHFFFAOYSA-N | |
5-(3-chlorophenyl)furan-2-carbonyl chloride | |
3854521 |
Chemical Identifiers
92973-26-7 | |
241.07 | |
DBONYOXUBWINEQ-UHFFFAOYSA-N | |
3854521 | |
ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1 |
C11H6Cl2O2 | |
MFCD02258034 | |
5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl | |
5-(3-chlorophenyl)furan-2-carbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures