missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Bromophenyl)isoxazole

$74.35 - $74.35

Chemical Identifiers

CAS 7064-33-7
Molecular Formula C9H6BrNO
Formula Weight 224.06
MDL Number MFCD02183531
Chemical Name or Material 5-(3-Bromophenyl)isoxazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50580MD SDS Alfa Aesar™
H50580MD
250mg Each for $74.35
AAH5058003 SDS Alfa Aesar™
H5058003
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7064-33-7
224.06
5-(3-Bromophenyl)isoxazole
C9H6BrNO
MFCD02183531

Specifications

7064-33-7
224.06
250mg
C9H6BrNO
MFCD02183531
5-(3-Bromophenyl)isoxazole