Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3-Bromophenyl)-1H-tetrazole, 97%

$67.05 - $185.34

Chemical Identifiers

Chemical Name or Material 5-(3-Bromophenyl)-1H-tetrazole
CAS 3440-99-1
Molecular Formula C7H5BrN4
Formula Weight 225.05
MDL Number MFCD01075675
Beilstein 141258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1518003

  SDS
alfa aesar™
L1518003
1g Each for $185.34

AAL15180MD

  SDS
alfa aesar™
L15180MD
250mg Each for $67.05
Description & Specifications

Specifications

Formula Weight 225.05
MDL Number MFCD01075675
Chemical Name or Material 5-(3-Bromophenyl)-1H-tetrazole
Molecular Formula C7H5BrN4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 3440-99-1
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 150° to 152°C
Beilstein 141258
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.