missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(3,4-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5415203

 View more versions of this product

Catalog No. AAH5415203



Specifications

Specifications

5-(3,4-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
1449142-43-1
MFCD20040143
QBXYLSUKPFQIGC-UHFFFAOYSA-N
[5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
73995661
97%
1g
C13H9BCl2FNO3
5-3,4-dichlorophenyl carbamoyl-2-fluorophenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC(=C(C=C2)Cl)Cl)F)(O)O
327.927
327.93

Chemical Identifiers

1449142-43-1
327.927
QBXYLSUKPFQIGC-UHFFFAOYSA-N
73995661
B(C1=C(C=CC(=C1)C(=O)NC2=CC(=C(C=C2)Cl)Cl)F)(O)O
C13H9BCl2FNO3
MFCD20040143
5-3,4-dichlorophenyl carbamoyl-2-fluorophenylboronic acid
[5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents