missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Thienyl)isoxazole-3-carboxylic acid, 95%

$56.59 - $168.09

Chemical Identifiers

CAS 763109-71-3
Molecular Formula C8H5NO3S
Molecular Weight (g/mol) 195.192
MDL Number MFCD04113955
InChI Key OBTRHBQUXRJHJR-UHFFFAOYSA-N
Synonym 5-thiophen-2-yl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-isoxazole-3-carboxylic acid, 5-2-thienyl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid, 5-thien-2-ylisoxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-thienyl, 5-2-thienyl-3-isoxazolecarboxylic acid, 5-2-thienyl isoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-2-thienyl, 5-thiophen-2-ylisoxazole-3-carboxylic acid
PubChem CID 3157098
IUPAC Name 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid
SMILES C1=CSC(=C1)C2=CC(=NO2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63824MD View Documents Alfa Aesar™
H63824MD
250mg Each for $56.59
 
AAH6382403 View Documents Alfa Aesar™
H6382403
1g Each for $168.09
 
Specifications

Chemical Identifiers

763109-71-3
195.192
OBTRHBQUXRJHJR-UHFFFAOYSA-N
3157098
C1=CSC(=C1)C2=CC(=NO2)C(=O)O
C8H5NO3S
MFCD04113955
5-thiophen-2-yl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-isoxazole-3-carboxylic acid, 5-2-thienyl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid, 5-thien-2-ylisoxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-thienyl, 5-2-thienyl-3-isoxazolecarboxylic acid, 5-2-thienyl isoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-2-thienyl, 5-thiophen-2-ylisoxazole-3-carboxylic acid
5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid

Specifications

250mg
C8H5NO3S
5-thiophen-2-yl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-isoxazole-3-carboxylic acid, 5-2-thienyl isoxazole-3-carboxylic acid, 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid, 5-thien-2-ylisoxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-thienyl, 5-2-thienyl-3-isoxazolecarboxylic acid, 5-2-thienyl isoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-2-thienyl, 5-thiophen-2-ylisoxazole-3-carboxylic acid
C1=CSC(=C1)C2=CC(=NO2)C(=O)O
195.192
195.2
95%
763109-71-3
MFCD04113955
OBTRHBQUXRJHJR-UHFFFAOYSA-N
5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid
3157098
Solid
5-(2-Thienyl)isoxazole-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning