Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Thienyl)-1H-pyrazole, 99%

$109.85 - $382.72

Chemical Identifiers

Chemical Name or Material 5-(2-Thienyl)-1H-pyrazole
CAS 19933-24-5
Molecular Formula C7H6N2S
Formula Weight 150.2
MDL Number MFCD00059740
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5065803

  SDS
alfa aesar™
H5065803
1g Each for $382.72

AAH50658MD

  SDS
alfa aesar™
H50658MD
250mg Each for $109.85
Description & Specifications

Specifications

Formula Weight 150.2
Chemical Name or Material 5-(2-Thienyl)-1H-pyrazole
CAS 19933-24-5
Assay 99%
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6N2S
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00059740
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.