missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(2-Pyridyl)thiophene-2-sulfonyl chloride, Technical Grade, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC02203DA
Specifications
5-(2-Pyridyl)thiophene-2-sulfonyl chloride | |
95% | |
MFCD00052284 | |
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride | |
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1 | |
259.72 | |
259.74 |
151858-64-9 | |
C9H6ClNO2S2 | |
1g | |
OGOMWPWAEIDEOU-UHFFFAOYSA-N | |
5-pyridin-2-ylthiophene-2-sulfonyl chloride | |
2737231 | |
95% |
Chemical Identifiers
151858-64-9 | |
259.72 | |
OGOMWPWAEIDEOU-UHFFFAOYSA-N | |
2737231 | |
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1 |
C9H6ClNO2S2 | |
MFCD00052284 | |
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride | |
5-pyridin-2-ylthiophene-2-sulfonyl chloride |