Please login to your online account to display your discounted pricing

5-(2-Pyridyl)thiophene-2-carboxylic acid, 97%


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC02201DA

 View more versions of this product

Catalog No. CC02201DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 205.24
Quantity 1g
Assay 97%
Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Assay Percent Range 97%
CAS 119082-97-2
Packaging Amber glass bottle
Molecular Formula C10H7NO2S

Specifications

Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7NO2S
Formula Weight 205.24