Please login to your online account to display your discounted pricing

5-(2-Pyridyl)thiophene-2-carboxylic acid, 97%


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC02201CB

 View more versions of this product

Catalog No. CC02201CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.24
Quantity 250mg
Assay 97%
Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C10H7NO2S
CAS 119082-97-2

Specifications

Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7NO2S
Formula Weight 205.24