Please login to your online account to display your discounted pricing

5-(2-Pyridyl)thiophene-2-carboxylic acid, 97%

$78.00 - $78.00

Chemical Identifiers

Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7NO2S
Formula Weight 205.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC02201CB

  SDS
fisher scientific company
CC02201CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC02201DA

  SDS
fisher scientific company
CC02201DA
Amber glass bottle 1g Each for $78.00

CC02201DE

 
fisher scientific company
CC02201DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.24
Quantity 250mg
Assay 97%
Chemical Name or Material 5-(2-Pyridyl)thiophene-2-carboxylic acid
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C10H7NO2S
CAS 119082-97-2