Please login to your online account to display your discounted pricing

5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Maybridge


Quantity 1g

Manufacturer: fisher scientific company  KM08625DA

 View more versions of this product

Catalog No. KM08625DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.2
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
CAS 130423-83-5
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)-2-furoic acid
MDL Number MFCD00541949
Molecular Formula C13H8O3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-(2-Phenyleth-1-ynyl)-2-furoic acid
CAS 130423-83-5
Assay Tech.
Assay Percent Range 95%
Molecular Formula C13H8O3
Formula Weight 212.2
MDL Number MFCD00541949
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents