missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Methyl-4-nitrophenyl)furan-2-carbonyl chloride

Chemical Identifiers

CAS 381178-60-5
Molecular Formula C12H8ClNO4
Formula Weight 265.65
MDL Number MFCD02257980
UN Number UN3261
Chemical Name or Material 5-(2-Methyl-4-nitrophenyl)furan-2-carbonyl chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5039203 SDS Alfa Aesar™
H5039203
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5039206 SDS Alfa Aesar™
H5039206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

381178-60-5
265.65
UN3261
C12H8ClNO4
MFCD02257980
5-(2-Methyl-4-nitrophenyl)furan-2-carbonyl chloride

Specifications

381178-60-5
265.65
UN3261
5-(2-Methyl-4-nitrophenyl)furan-2-carbonyl chloride
C12H8ClNO4
MFCD02257980
1g

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.