Please login to your online account to display your discounted pricing

5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18203DE

 View more versions of this product

Catalog No. CC18203DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 279.79
Quantity 5g
Assay 95%
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C8H6ClNO2S3
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride
CAS 215434-25-6

Specifications

Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6ClNO2S3
Formula Weight 279.79