missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC18203CB
Specifications
5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride | |
95% | |
C8H6ClNO2S3 | |
5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone | |
CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl | |
279.771 | |
279.79 |
215434-25-6 | |
Amber Glass Bottle | |
250 mg | |
YIKNRCVJBOLZFL-UHFFFAOYSA-N | |
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride | |
2736979 | |
95% |
Chemical Identifiers
215434-25-6 | |
279.771 | |
5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone | |
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride |
C8H6ClNO2S3 | |
YIKNRCVJBOLZFL-UHFFFAOYSA-N | |
2736979 | |
CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl |