missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18201DA

 View more versions of this product

Catalog No. CC18201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarboxylic acid
96.5
97%
C9H7NO2S2
1g
AEDWMWQOFJYAGN-UHFFFAOYSA-N
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
614467
97%
400715-45-9
100.0
Amber glass bottle
MFCD02682053
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
CC1=NC(=CS1)C2=CC=C(S2)C(=O)O
225.28
225.29

Chemical Identifiers

400715-45-9
225.28
AEDWMWQOFJYAGN-UHFFFAOYSA-N
614467
CC1=NC(=CS1)C2=CC=C(S2)C(=O)O
C9H7NO2S2
MFCD02682053
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions