missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarbaldehyde, Maybridge™
Chemical Identifiers
CAS |
499770-66-0 |
Molecular Formula |
C9H7NOS2 |
Molecular Weight (g/mol) |
209.281 |
MDL Number |
MFCD02682090 |
InChI Key |
OTEBKLSFEFZNAG-UHFFFAOYSA-N |
Synonym |
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carbaldehyde, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxaldehyde, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarbaldehyde, 5-2-methylthiazol-4-yl thiophene-2-carbaldehyde |
PubChem CID |
2776469 |
IUPAC Name |
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carbaldehyde |
SMILES |
CC1=NC(=CS1)C2=CC=C(S2)C=O |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
499770-66-0 |
|
209.281 |
|
OTEBKLSFEFZNAG-UHFFFAOYSA-N |
|
2776469 |
|
CC1=NC(=CS1)C2=CC=C(S2)C=O |
|
C9H7NOS2 |
|
MFCD02682090 |
|
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carbaldehyde, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxaldehyde, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarbaldehyde, 5-2-methylthiazol-4-yl thiophene-2-carbaldehyde |
|
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carbaldehyde |
Specifications
|
Amber glass bottle |
|
499770-66-0 |
|
100.0 |
|
MFCD02682090 |
|
OTEBKLSFEFZNAG-UHFFFAOYSA-N |
|
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carbaldehyde |
|
2776469 |
|
Tech. |
|
5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarbaldehyde |
|
250mg |
|
80.0 |
|
C9H7NOS2 |
|
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carbaldehyde, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxaldehyde, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarbaldehyde, 5-2-methylthiazol-4-yl thiophene-2-carbaldehyde |
|
CC1=NC(=CS1)C2=CC=C(S2)C=O |
|
209.281 |
|
209.29 |
|
Technical |
SDS