Please login to your online account to display your discounted pricing

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarbaldehyde, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC18204DA

 View more versions of this product

Catalog No. CC18204DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 209.29
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarbaldehyde
MDL Number MFCD02682090
CAS 499770-66-0
Packaging Amber glass bottle
Molecular Formula C9H7NOS2

Specifications

Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarbaldehyde
CAS 499770-66-0
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Molecular Formula C9H7NOS2
Formula Weight 209.29
MDL Number MFCD02682090