missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Maybridge

$41.39 - $171.25

Chemical Identifiers

Assay Percent Range 95%
Molecular Formula C9H9NOS2
Formula Weight 211.31
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC18209EB Fisher Scientific Company
CC18209EB
Amber Glass Bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18209DE Fisher Scientific Company
CC18209DE
Amber Glass Bottle 5g Each for $171.25
CC18209DA SDS Fisher Scientific Company
CC18209DA
1g Each for $41.39
Specifications Description & Specifications

Chemical Identifiers

95%
211.31
95%
C9H9NOS2
[5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol

Specifications

211.31
Amber Glass Bottle
95%
[5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
25g
C9H9NOS2
337508-70-0
95%