Please login to your online account to display your discounted pricing

5-(2-Furyl)pyridine-2-carbaldehyde, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC55104CB

 View more versions of this product

Catalog No. CC55104CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.17
Quantity 250mg
Assay 95%
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H7NO2
CAS 886851-45-2
Chemical Name or Material 5-(2-Furyl)pyridine-2-carbaldehyde
Packaging Amber Glass Bottle
MDL Number MFCD10700053
CAS Min % 94.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-(2-Furyl)pyridine-2-carbaldehyde
CAS 886851-45-2
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD10700053
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.