missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[5-(2-Furyl)isoxazol-3-yl]methylamine, ≥97%, Maybridge

Chemical Identifiers

CAS 850375-13-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H8N2O2
Formula Weight 164.16
MDL Number MFCD07368533
Chemical Name or Material [5-(2-Furyl)isoxazol-3-yl]methylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC34913DE Fisher Scientific Company
CC34913DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34913CB SDS Fisher Scientific Company
CC34913CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34913DA SDS Fisher Scientific Company
CC34913DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

850375-13-2
100.0
C8H8N2O2
MFCD07368533
97%
96.5
97%
164.16
[5-(2-Furyl)isoxazol-3-yl]methylamine

Specifications

850375-13-2
100.0
C8H8N2O2
MFCD07368533
[5-(2-Furyl)isoxazol-3-yl]methylamine
96.5
97%
164.16
5g
97%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.