Please login to your online account to display your discounted pricing

5-(2-Furyl)cyclohexane-1,3-dione, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CD11333DE

 View more versions of this product

Catalog No. CD11333DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 178.19
Quantity 5g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 5-(2-Furyl)cyclohexane-1, 3-dione
Molecular Formula C10H10O3
MDL Number MFCD00104796
CAS 1774-11-4

Specifications

Chemical Name or Material 5-(2-Furyl)cyclohexane-1, 3-dione
CAS 1774-11-4
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00104796