missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
5-(2-Furyl)cyclohexane-1,3-dione, 97%, Maybridge™
Chemical Identifiers
CAS |
1774-11-4 |
Molecular Formula |
C10H10O3 |
Molecular Weight (g/mol) |
178.187 |
MDL Number |
MFCD00104796 |
InChI Key |
FYLTVHCMIYGVPZ-UHFFFAOYSA-N |
Synonym |
1,3-cyclohexanedione, 5-2-furanyl, 1,3-cyclohexanedione,5-2-furanyl, 5-2-furanyl cyclohexane-1,3-dione, 5-2-furyl cyclohexan-1,3-dione, 5-2-furyl cyclohexane-1,3-dione, 5-2-furyl-1,3-cyclohexanedione, 5-2-furyl-cyclohexane-1,3-dione, 5-furan-2-yl cyclohexane-1,3-dione, 5-furan-2-yl-cyclohexane-1,3-dione, maybridge1_003103 |
PubChem CID |
699513 |
IUPAC Name |
5-(furan-2-yl)cyclohexane-1,3-dione |
SMILES |
C1C(CC(=O)CC1=O)C2=CC=CO2 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
1774-11-4 |
|
178.187 |
|
FYLTVHCMIYGVPZ-UHFFFAOYSA-N |
|
699513 |
|
C1C(CC(=O)CC1=O)C2=CC=CO2 |
|
C10H10O3 |
|
MFCD00104796 |
|
1,3-cyclohexanedione, 5-2-furanyl, 1,3-cyclohexanedione,5-2-furanyl, 5-2-furanyl cyclohexane-1,3-dione, 5-2-furyl cyclohexan-1,3-dione, 5-2-furyl cyclohexane-1,3-dione, 5-2-furyl-1,3-cyclohexanedione, 5-2-furyl-cyclohexane-1,3-dione, 5-furan-2-yl cyclohexane-1,3-dione, 5-furan-2-yl-cyclohexane-1,3-dione, maybridge1_003103 |
|
5-(furan-2-yl)cyclohexane-1,3-dione |
Specifications
|
Amber Glass Bottle |
|
1774-11-4 |
|
C10H10O3 |
|
1,3-cyclohexanedione, 5-2-furanyl, 1,3-cyclohexanedione,5-2-furanyl, 5-2-furanyl cyclohexane-1,3-dione, 5-2-furyl cyclohexan-1,3-dione, 5-2-furyl cyclohexane-1,3-dione, 5-2-furyl-1,3-cyclohexanedione, 5-2-furyl-cyclohexane-1,3-dione, 5-furan-2-yl cyclohexane-1,3-dione, 5-furan-2-yl-cyclohexane-1,3-dione, maybridge1_003103 |
|
C1C(CC(=O)CC1=O)C2=CC=CO2 |
|
178.187 |
|
178.19 |
|
5-(2-Furyl)cyclohexane-1, 3-dione |
|
1g |
|
97% |
|
MFCD00104796 |
|
FYLTVHCMIYGVPZ-UHFFFAOYSA-N |
|
5-(furan-2-yl)cyclohexane-1,3-dione |
|
699513 |
|
97% |
SDS