Please login to your online account to display your discounted pricing

[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material [5-(2-Furyl)-1H-pyrazol-3-yl]methanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2O2
Formula Weight 164.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC35009DE

 
fisher scientific company
CC35009DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC35009DA

  SDS
fisher scientific company
CC35009DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC35009CB

  SDS
fisher scientific company
CC35009CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.16
Quantity 5g
Assay 97%
Chemical Name or Material [5-(2-Furyl)-1H-pyrazol-3-yl]methanol
Assay Percent Range 97%
Packaging Amber Glass Bottle
CAS 84978-67-6
Molecular Formula C8H8N2O2