missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC35009DA
Specifications
[5-(2-Furyl)-1H-pyrazol-3-yl]methanol | |
97% | |
C8H8N2O2 | |
5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol | |
C1=COC(=C1)C2=NNC(=C2)CO | |
164.164 | |
164.16 |
84978-67-6 | |
Amber Glass Bottle | |
1g | |
NBADTAIDQSLLHA-UHFFFAOYSA-N | |
[3-(furan-2-yl)-1H-pyrazol-5-yl]methanol | |
7060534 | |
97% |
Chemical Identifiers
84978-67-6 | |
164.164 | |
5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol | |
[3-(furan-2-yl)-1H-pyrazol-5-yl]methanol |
C8H8N2O2 | |
NBADTAIDQSLLHA-UHFFFAOYSA-N | |
7060534 | |
C1=COC(=C1)C2=NNC(=C2)CO |