missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid, ≥97%, Maybridge

Chemical Identifiers

CAS 108128-39-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8N2O3
Formula Weight 192.17
MDL Number MFCD08060535
Chemical Name or Material 5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC46901CB SDS Fisher Scientific Company
CC46901CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC46901DA SDS Fisher Scientific Company
CC46901DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108128-39-8
100.0
C9H8N2O3
MFCD08060535
97%
96.5
97%
192.17
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid

Specifications

108128-39-8
100.0
Amber Glass Bottle
192.17
250mg
97%
96.5
97%
C9H8N2O3
MFCD08060535
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.