missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride, 97%, Maybridge

Chemical Identifiers

CAS 876316-47-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H7ClN2O2
Formula Weight 210.62
MDL Number MFCD08271946
Chemical Name or Material 5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC46902DA SDS Fisher Scientific Company
CC46902DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC46902CB SDS Fisher Scientific Company
CC46902CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

876316-47-1
100.0
C9H7ClN2O2
MFCD08271946
97%
96.5
95%
210.62
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride

Specifications

876316-47-1
100.0
Amber Glass Bottle
210.62
1g
97%
96.5
95%
C9H7ClN2O2
MFCD08271946
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger