missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbaldehyde, ≥97%, Maybridge

Chemical Identifiers

CAS 876728-40-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8N2O2
Formula Weight 176.17
MDL Number MFCD08271947
Chemical Name or Material 5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC46904CB SDS Fisher Scientific Company
CC46904CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC46904DA SDS Fisher Scientific Company
CC46904DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

876728-40-4
100.0
C9H8N2O2
MFCD08271947
97%
96.5
97%
176.17
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbaldehyde

Specifications

176.17
100.0
876728-40-4
C9H8N2O2
5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbaldehyde
250mg
MFCD08271947
96.5
97%
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.