Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Fluorophenyl)-1H-tetrazole, 95%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material 5-(2-Fluorophenyl)-1H-tetrazole
CAS 50907-19-2
Molecular Formula C7H5FN4
Formula Weight 164.14
MDL Number MFCD00798373
Beilstein 1105868
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1925903

  SDS
alfa aesar™
L1925903
1g Each for $226.01
Description & Specifications

Specifications

Formula Weight 164.14
Molecular Formula C7H5FN4
Health Hazard 1 Warning
Chemical Name or Material 5-(2-Fluorophenyl)-1H-tetrazole
Assay 95%
CAS 50907-19-2
Beilstein 1105868
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00798373
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 157° to 159°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.