missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

$180.00 - $573.77

Chemical Identifiers

CAS 1449144-63-1
Molecular Formula C13H10BClFNO3
Molecular Weight (g/mol) 293.485
MDL Number MFCD20040195
InChI Key XLAINFWLAVXHRA-UHFFFAOYSA-N
Synonym 5-2-chlorophenyl carbamoyl-2-fluorophenylboronic acid
PubChem CID 73995825
IUPAC Name [5-[(2-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2Cl)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54470MD View Documents Alfa Aesar™
H54470MD
250mg Each for $180.00
 
AAH5447003 View Documents Alfa Aesar™
H5447003
1g Each for $573.77
 
Specifications

Chemical Identifiers

1449144-63-1
293.485
XLAINFWLAVXHRA-UHFFFAOYSA-N
73995825
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2Cl)F)(O)O
C13H10BClFNO3
MFCD20040195
5-2-chlorophenyl carbamoyl-2-fluorophenylboronic acid
[5-[(2-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid

Specifications

250mg
C13H10BClFNO3
5-2-chlorophenyl carbamoyl-2-fluorophenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2Cl)F)(O)O
293.485
293.48
5-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
1449144-63-1
MFCD20040195
XLAINFWLAVXHRA-UHFFFAOYSA-N
[5-[(2-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
73995825
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures