missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5447003

 View more versions of this product

Catalog No. AAH5447003



Specifications

Specifications

5-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
1449144-63-1
MFCD20040195
XLAINFWLAVXHRA-UHFFFAOYSA-N
[5-[(2-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
73995825
97%
1g
C13H10BClFNO3
5-2-chlorophenyl carbamoyl-2-fluorophenylboronic acid, 5-2-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-2-chlorophenylcarbamoyl-2-fluorophenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2Cl)F)(O)O
293.485
293.48

Chemical Identifiers

1449144-63-1
293.485
XLAINFWLAVXHRA-UHFFFAOYSA-N
73995825
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2Cl)F)(O)O
C13H10BClFNO3
MFCD20040195
5-2-chlorophenyl carbamoyl-2-fluorophenylboronic acid, 5-2-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-2-chlorophenylcarbamoyl-2-fluorophenylboronic acid
[5-[(2-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents