Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chlorophenyl)-1H-tetrazole, 98%

$64.19 - $233.97

Chemical Identifiers

Chemical Name or Material 5-(2-Chlorophenyl)-1H-tetrazole
CAS 50907-46-5
Molecular Formula C7H5ClN4
Formula Weight 180.6
MDL Number MFCD00040950
Beilstein 148559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1116403

 
alfa aesar™
L1116403
1g Each for $64.19

AAL1116406

 
alfa aesar™
L1116406
5g Each for $233.97
Description & Specifications

Specifications

Formula Weight 180.6
Chemical Name or Material 5-(2-Chlorophenyl)-1H-tetrazole
Assay 98%
Molecular Formula C7H5ClN4
MDL Number MFCD00040950
CAS 50907-46-5
Beilstein 148559
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 179° to 180°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.