missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chlorophenyl)-1H-tetrazole, 98%

Manufacturer:  Alfa Aesar™ L1116403

 View more versions of this product

Catalog No. AAL1116403



Specifications

Specifications

5-(2-Chlorophenyl)-1H-tetrazole
1g
C7H5ClN4
148559
PSUIIKIEUATWCZ-UHFFFAOYSA-N
5-(2-chlorophenyl)-2H-tetrazole
560735
98%
179°C to 180°C
50907-46-5
MFCD00040950
5-2-chlorophenyl-1h-tetrazole, 5-2-chlorophenyl-2h-tetrazole, 5-2-chlorophenyl-2h-1,2,3,4-tetrazole, 5-2-chlorophenyl tetrazole, 5-2-chlorophenyl-2h-tetraazole, 2h-tetrazole,5-2-chlorophenyl, 5-2-chlorophenyl-1h-1,2,3,4-tetrazole, 5-2-chlorophenyl-1h-1,2,3,4-tetraazole, ldha inhibitor, 14, 2-chlorophenyl tetrazole
C1=CC=C(C(=C1)C2=NNN=N2)Cl
180.595
180.6

Chemical Identifiers

50907-46-5
180.595
PSUIIKIEUATWCZ-UHFFFAOYSA-N
560735
C1=CC=C(C(=C1)C2=NNN=N2)Cl
C7H5ClN4
MFCD00040950
5-2-chlorophenyl-1h-tetrazole, 5-2-chlorophenyl-2h-tetrazole, 5-2-chlorophenyl-2h-1,2,3,4-tetrazole, 5-2-chlorophenyl tetrazole, 5-2-chlorophenyl-2h-tetraazole, 2h-tetrazole,5-2-chlorophenyl, 5-2-chlorophenyl-1h-1,2,3,4-tetrazole, 5-2-chlorophenyl-1h-1,2,3,4-tetraazole, ldha inhibitor, 14, 2-chlorophenyl tetrazole
5-(2-chlorophenyl)-2H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents