missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%

$136.97 - $436.24

Chemical Identifiers

CAS 1449133-18-9
Molecular Formula C13H9BClFN2O5
Molecular Weight (g/mol) 338.482
MDL Number MFCD20126195
InChI Key SMRJKQIAHFDGKT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-nitrophenyl carbamoyl-5-fluorophenylboronic acid
PubChem CID 73995623
IUPAC Name [3-[(2-chloro-6-nitrophenyl)carbamoyl]-5-fluorophenyl]boronic acid
SMILES B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=CC=C2Cl)[N+](=O)[O-])(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54080MD View Documents Alfa Aesar™
H54080MD
250mg Each for $136.97
 
AAH5408003 View Documents Alfa Aesar™
H5408003
1g Each for $436.24
 
Specifications

Chemical Identifiers

1449133-18-9
338.482
SMRJKQIAHFDGKT-UHFFFAOYSA-N
73995623
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=CC=C2Cl)[N+](=O)[O-])(O)O
C13H9BClFN2O5
MFCD20126195
3-2-chloro-6-nitrophenyl carbamoyl-5-fluorophenylboronic acid
[3-[(2-chloro-6-nitrophenyl)carbamoyl]-5-fluorophenyl]boronic acid

Specifications

250mg
C13H9BClFN2O5
3-2-chloro-6-nitrophenyl carbamoyl-5-fluorophenylboronic acid
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=CC=C2Cl)[N+](=O)[O-])(O)O
338.482
338.49
5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid
1449133-18-9
MFCD20126195
SMRJKQIAHFDGKT-UHFFFAOYSA-N
[3-[(2-chloro-6-nitrophenyl)carbamoyl]-5-fluorophenyl]boronic acid
73995623
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures