Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chloro-6-fluorobenzyl)-2H-tetrazole

$164.51 - $164.51

Chemical Identifiers

Chemical Name or Material 5-(2-Chloro-6-fluorobenzyl)-2H-tetrazole
CAS 874607-03-1
Molecular Formula C8H6ClFN4
Formula Weight 212.62g/mol
MDL Number MFCD00973901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5043603

 
alfa aesar™
H50436-03
Each for $164.51
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 5-(2-Chloro-6-fluorobenzyl)-2H-tetrazole
CAS 874607-03-1
Molecular Formula C8H6ClFN4
Formula Weight 212.62g/mol
MDL Number MFCD00973901