Please login to your online account to display your discounted pricing

5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
CAS 306936-04-9
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H6ClF3O2
Formula Weight 274.62
MDL Number MFCD01571128
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW06174DA

 
fisher scientific company
SEW06174DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.62
Quantity 1g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Chemical Name or Material 5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
Packaging Amber glass bottle
Molecular Formula C12H6ClF3O2
MDL Number MFCD01571128
CAS 306936-04-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.